Thermo Scientific Orion Star A100 Series Benchtop pH/mV/Temperature Meter
Make it easy and affordable to take simple, accurate pH measurements
  • Large LCD displays pH or mV along with temperature
  • Up to 3-point pH calibration, with easy recall of slope data
  • Auto-Read™ function locks in a stable reading
  • Automatic recognition of USA, NIST, and DIN buffers
  • Nonvolatile memory holds up to 50 data points
  • Swing-arm electrode holder stand features a newly designed probe holder
  • Option of AC power or four AA batteries (sold separately)
Thermo Scientific Orion Star A111 benchtop pH meter combines simplicity with accuracy. Large informative display shows readings, battery life, electrode status, meter mode, calibration information, power source and more. Simple button layout and on-screen messages help with calibration and setup menu choices.
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Thermo Scientific STAR A111 pH Benchtop Meter Only EW-58825-02 Mfr # STARA1110
COMPARE
Star A111 pH Benchtop Meter Only
$587.47USD / Each
$652.75USD / Each
Thermo Scientific STAR A111 Star A111 pH Benchtop Meter Kit EW-58825-04 Mfr # STARA1115
COMPARE
Star A111 pH Benchtop Meter Kit
$877.72USD / Each
$975.25USD / Each