Thermo Scientific Orion Star A 300 pH and pH/Ion Meters
Advanced features and dependability makes it the top choice for measuring samples
  • Multilanguage interface supports Engliah, Spanish, French, Italian, German and Chinese
  • Improved stability algorithm provide better measurement accuracy
  • Ensure temperature accuracy with ATC temperature calibration
  • Nonvolatile memory holds up to 5000 data points with time date stamp
  • Individual method passwords provide better security
  • Four AA batteries provide over 800 hours of field use, or order optional AC adapter for lab use
Thermo Scientific Orion Star A 300 meters features an intuitive user interface with plain language screen prompts. A large, backlit graphic LCD, shows measurement and temperature along with electrode status, time/date, sample ID, user ID, and calibration data.
GLP-related features include 5000 data points with time/date stamp and electrode verification. Meter utilizes RS-232 or USB for connectivity and includes data analysis software.
ITEM(S) ON SALE
$1,141.20 - $2,160.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Max ISE Max pH (pH) Description
Availability Pricing
Thermo Scientific A321 pH Portable Meter Only EW-58825-44 Mfr # STARA3210
COMPARE
20 pH Portable Meter Only
$1,141.20USD / Each
$1,268.00USD / Each
Thermo Scientific A321 pH Portable Meter Kit EW-58825-46 Mfr # STARA3215
COMPARE
20 pH Portable Meter Kit
$1,878.30USD / Each
$2,087.00USD / Each
Thermo Scientific A324 pH/ISE Portable Meter Only EW-58825-64 Mfr # STARA3240
COMPARE
19999 ppm 20 pH/ISE Portable Meter Only
$1,586.70USD / Each
$1,763.00USD / Each
Thermo Scientific A324 pH/ISE Portable Meter Kit EW-58825-66 Mfr # STARA3245
COMPARE
19999 ppm 20 pH/ISE Portable Meter Kit
$2,160.00USD / Each
$2,400.00USD / Each