Thermo Scientific Orion High-Performance Ammonia Electrode
Designed to meet the demanding needs of both drinking water and wastewater applications
  • Compliant with EPA testing method for ammonia measurement
  • Saves time to help make your lab more efficient
This Orion high performance ammonia electrode is extremely durable with a chemical-resistant translucent body. The easy-to-fill electrode comes marked with a fill line to avoid overfilling and allows you to monitor the fill solution level without disassembling the electrode. Membrane replacement options include cost-saving loose membranes or preassembled electrode body with membrane for the convenience of not having to install your own membrane.
$1,115.00 - $1,167.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Electrode Body Material Electrode Type
Availability Pricing
Thermo Scientific 9512BNWP ISE Ammonia Electrode with BNC Connector EW-05722-16 Mfr # 9512BNWP
COMPARE
Epoxy Gas sensing
$1,115.00USD / Each
Thermo Scientific 9512HPBNWP ISE electrode, Hi-Performance Ammonia, BNC EW-05722-18 Mfr # 9512HPBNWP
COMPARE
Translucent plastic Gas sensing
$1,167.00USD / Each