Thermo Scientific Orion Dual Star Benchtop pH/ISE/mV Meter
Innovative EZ Startup™ ensures no manual is needed to begin taking measurements
  • Dual inputs allow you to simultaneously measure and display pH, ISE, and temperature
  • Simple to operate — prompts walk you through meter operation and setup
The Orion Dual Star meter provides dual BNC and temperature inputs for simultaneous pH and ISE readings. Ion determination is facilitated by additional options for concentration units, optional auto-blank for low-level measurements, built-in Star-stirrer control, and the ability to view, print your calibration graph with slope data, and modify calibration points. The meter easily guides you through common direct measurements, but also is programmed for a range of single and double known addition and subtraction methods. Autosampler capabilities allow you to take advantage of high sample throughput technology to save time and improve measurement quality.
The Dual Star utilizes EZ Startup™ for setup without the manual, and Smart Stability™ and SmartAveraging™ to ensure accurate results. The meter offers a full complement of GLP-related features including 1000 point datalogging with time/date stamp, electrode verification, and full calibration data. Generate and store up to 10 password-protected methods per channel. Easily download from the USB port or bidirectional RS-232.
Meter includes: Quick-Start guide, electrode stand, stainless steel ATC probe, and universal AC power supply.
Meter with electrode adds: a refillable, glass-body Orion ROSS Ultra® electrode.
Meter kit adds: 475-mL bottles of 4, 7, and 10 buffer; and Orion ROSS® electrode storage solution.
ITEM(S) ON SALE
$2,212.20 - $2,646.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max mV (mV) Max pH (pH) Description
Availability Pricing
Thermo Scientific Dual Star pH/ISE/mV Meter Only EW-58825-00 Mfr # 2115000
COMPARE
1999 20 pH/ISE/mV Meter Only
$2,212.20USD / Each
$2,458.00USD / Each
Thermo Scientific Dual Star pH/ISE/mV Meter with Electrode EW-58825-10 Mfr # 2115101
COMPARE
1999 20 pH/ISE/mV Meter with Electrode
$2,608.20USD / Each
$2,898.00USD / Each
Thermo Scientific Dual Star pH/ISE/mV Meter Kit EW-58825-20 Mfr # 2115001
COMPARE
1999 20 pH/ISE/mV Meter Kit
$2,646.00USD / Each
$2,940.00USD / Each