Thermo Scientific™ Orion™ Combination Ion-Selective Electrodes
Orion-designed for optimum performance and the most comprehensive ISE selection
  • ionplus™ Sure-Flow™ Combination Ion-Selective Electrodes eliminate fumbling and tangles from multiple cables
  • Combination Ion-Selective Electrodes feature both half-cell and reference cell in one electrode
  • Half-Cell Ion-Selective Electrodes are the classic means of taking ion measurements
Combination Ion-Selective Electrodes are easier to use than reference and half-cells—eliminating fumbling and tangles from multiple cables. Sure-flow® technology provides stable, reproducible results. The easy-to-clean design reduces the risk of inaccurate readings due to clogging of the reference junction. All electrodes include a BNC connector, a 3-ft cable, and manual. Complete systems require-the purchase of a standardizing solution, filling solution for the reference electrode, ionic strength adjustor, and pH adjusters. Chloride, cyanide, Sulfide, nitrate, and fluoride are approved for EPA methods for analysis of wastewater and for NPDES permitting.
ITEM(S) ON SALE
$1,060.00 - $1,566.00USD / Each
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Electrode Type Ion-Selective Electrode
Availability Pricing
Thermo Scientific 9635BNWP Sure-Flow Ion-Selective Electrode; Bromide EW-05713-01 Mfr # 9635BNWP
COMPARE
Solid-state Bromide
$1,339.00USD / Each
Thermo Scientific 9629BNWP Sure-Flow Ion-Selective Electrode; Cupric EW-05713-07 Mfr # 9629BNWP
COMPARE
Solid-state Copper
$1,566.00USD / Each
Thermo Scientific 9606BNWP Sure-Flow Ion-Selective Electrode; Cyanide EW-05713-09 Mfr # 9606BNWP
COMPARE
Solid-state Cyanide
$1,244.00USD / Each
Thermo Scientific 9653BNWP Sure-Flow Ion-Selective Electrode; Iodide EW-05713-11 Mfr # 9653BNWP
COMPARE
Solid-state Iodide
$1,270.00USD / Each
Thermo Scientific 9707BNWP Sure-Flow Ion-Selective Electrode; Nitrate EW-05713-15 Mfr # 9707BNWP
COMPARE
Membrane Nitrate
$1,464.00USD / Each
Thermo Scientific 9616BNWP Sure-Flow Ion-Selective Electrode; Silver/Sulfer EW-05713-21 Mfr # 9616BNWP
COMPARE
Solid-state Silver/Sulfide
$1,411.00USD / Each
Thermo Scientific 9502BNWP ISE Electrode; Carbon Dioxide/Carbonate EW-05722-01 Mfr # 9502BNWP
COMPARE
Membrane Carbon dioxide/Carbonate
$1,315.00USD / Each
Thermo Scientific 9609BNWP Sure-Flow Ion-Selective Electrode; Fluoride EW-05723-02 Mfr # 9609BNWP
COMPARE
Solid-state Fluoride
$1,210.00USD / Each
Thermo Scientific 8611BNWP ISE Electrode; Sodium EW-05723-31 Mfr # 8611BNWP
COMPARE
Solid-state Sodium
$1,060.00USD / Each
Thermo Scientific 9770BNWP ISE Electrode; Chlorine EW-05724-02 Mfr # 9770BNWP
COMPARE
Solid-state Residual Chlorine
$1,092.00USD / Each
Thermo Scientific 9720BNWP Sure-Flow Ion-Selective Electrode; Calcium EW-58824-52 Mfr # 9720BNWP
COMPARE
Membrane Calcium
$1,200.60USD / Each
$1,334.00USD / Each
Thermo Scientific 9719BNWP Sure-Flow Ion-Selective Electrode; Potassium EW-58824-56 Mfr # 9719BNWP
COMPARE
Membrane Potassium
$1,343.70USD / Each
$1,493.00USD / Each
Thermo Scientific 9617BNWP Sure-Flow Ion-Selective Electrode; Chloride EW-58824-60 Mfr # 9617BNWP
COMPARE
Solid-state Chloride
$1,129.50USD / Each
$1,255.00USD / Each
Showing 1- 13 of 13