Thermo Scientific Nunc CryoTube Internal Thread Cryogenic Vials
  • Polyethylene closure with silicone gasket provides a secure seal.
Presterilized by cobalt-60 radiation, these one-piece molded polypropylene vials withstand temperatures from -196 to 135°C. Starfooted vials lock into CryoTube® vial holder 03755-71 for one-handed operation. Optional colored cap inserts simplify sample identification.
Note: Do not autoclave or immerse vials in liquid nitrogen. For storage under extreme conditions such as liquid nitrogen, use CryoFlex™ tubing 03755-64.
$49.00 - $131.00USD / Bag of 50
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tube Bottom Shape Volume (mL)
Availability Pricing
Thermo Scientific Nunc 368632 CryoTube, free standing round bottom; 1.8 mL, 50/bag
Buy More Save More
EW-03755-10 Mfr # 368632
COMPARE
Round 1.8
$55.00USD / Bag of 50
Thermo Scientific Nunc 363401 CryoTube cryogenic tube with round bottom; 1.8 mL, 50/bag
Buy More Save More
EW-03755-20 Mfr # 363401
COMPARE
Round 1.8
$49.00USD / Bag of 50
Thermo Scientific Nunc 366656 CryoTubes, Internal Thread/Conical/1.0 mL; 50/Bag
Buy More Save More
EW-03755-30 Mfr # 366656
COMPARE
Conical 1
$49.00USD / Bag of 50
Thermo Scientific Nunc 363452 CryoTube cryogenic tube with round bottom; 4.5 mL, 50/bag
Buy More Save More
EW-03755-40 Mfr # 363452
COMPARE
Round 4.5
$73.00USD / Bag of 50
Thermo Scientific Nunc 366524 CryoTubes, Internal Thread/Round/3.6 mL; 50/Bag
Buy More Save More
EW-03755-50 Mfr # 366524
COMPARE
Round 3.6
$124.00USD / Bag of 50
Thermo Scientific Nunc 377224 CryoTubes, Internal Thread/Starfoot/1.0 mL; 50/Pk EW-03757-60 Mfr # 377224
COMPARE
Conical 1
$49.00USD / Pkg of 50
Thermo Scientific Nunc 377267 CryoTubes, Internal Thread/Starfoot/1.8 mL; 50/Pk EW-03757-62 Mfr # 377267
COMPARE
Round 1.8
$53.00USD / Pkg of 50
Thermo Scientific Nunc 379189 CryoTubes, Internal Thread/Starfoot/3.6 mL; 100/Pk EW-03757-64 Mfr # 379189
COMPARE
Starfoot 3.6
$131.00USD / Pkg of 50
Thermo Scientific Nunc 379146 CryoTubes, Internal Thread/Starfoot/4.5 mL; 50/Pk EW-03757-66 Mfr # 379146
COMPARE
Round 4.5
$77.00USD / Pkg of 50
Showing 1- 9 of 9