Thermo Scientific™ Nalgene™ Cryogenic Vials, External Thread, Sterile
External threads and long skirt length reduce risk of contamination
  • Polypropylene vials with HDPE closures are self-standing
  • Vials withstand temperatures from –196 to 120°C
White marking area aids in sample identification. Graduated on the side. Order colored cap inserts separately.
Note: Do not autoclave or immerse vials in liquid nitrogen. For storage under extreme conditions such as liquid nitrogen, use CryoFlex™ tubing 03755-64 (order separately).
$26.00 - $38.50USD / Pkg of 10
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (mm) Outside Diameter (mm) Volume (mL)
Availability Pricing
Thermo Scientific Nalgene Cryogenic Vial, Sterile, Polypropylene, External Thread, 1 mL; 25/Pk EW-06754-93 Mfr # 5000-1012
COMPARE
39 12 1
$38.50USD / Pkg of 25
Thermo Scientific Nalgene Cryogenic Vial, Sterile, Polypropylene, External Thread, 1.5 mL; 25/Pk
Always in Stock
EW-06754-94 Mfr # 5000-1020
COMPARE
48 12 1.5
$38.50USD / Pkg of 25
Thermo Scientific Nalgene Cryogenic Vial, Sterile, Polypropylene, External Thread, 1.2 mL; 25/Pk EW-06754-95 Mfr # 5000-0012
COMPARE
38 13.5 1.2
$32.50USD / Pkg of 25
Thermo Scientific Nalgene Cryogenic Vial, Sterile, Polypropylene, External Thread, 2 mL; 25/Pk
Always in Stock
EW-06754-96 Mfr # 5000-0020
COMPARE
48 13.5 2
$35.00USD / Pkg of 25
Thermo Scientific Nalgene Cryogenic Vial, Sterile, Polypropylene, External Thread, 5 mL; 10/Pk EW-06754-97 Mfr # 5000-0050
COMPARE
89.4 13.5 5
$26.00USD / Pkg of 10