Thermo Scientific™ GENESYS™ Visible and UV-Visible Spectrophotometers
Accurate effortless quantitative analysis
  • Large sample compartment with interchangeable holders increase versatility
  • SmartStart™ menu stores programs for quick recall of routine methods
  • Easy reporting via USB data export or optional snap-on printer
  • Simple prealigned lamp replacement reduces mistakes and downtime
Thermo GENESYS spectrophotometers feature simple-to-use, high resolution, color display start menus allowing you to start making measurements immediately. Expand capabilities further with optional VISIONlite™ software (order separately), which provides unlimited method storage and advanced modes for scanning, quantitative analysis, and kinetics applications.
Enhanced flexibility features include interchangeable sample holders, uncomplicated lamp and lid replacement, and multiple data transfer options. Choose from single, dual, or double beam optics for a wide range of applications.
$3,634.00 - $8,827.00USD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Bandwidth Beam Type Min Wavelength (nm)
Availability Pricing
Thermo Scientific Genesys GENESYS™ 30  GENESYS™ 30 Visible Light Spectrophotometer, US plug EW-02654-01 Mfr # 840-277000
COMPARE
5 nm Single beam 325
$3,634.00USD / Each
Thermo Scientific GENESYS™ 40 GENESYS™ 40 Visible Light Spectrophotometer, US plug EW-02654-24 Mfr # 840-297000
COMPARE
5 nm Dual beam 325
$5,075.00USD / Each
Thermo Scientific GENESYS™ 50 GENESYS™ 50 UV-Visible Light Spectrophotometer, US plug EW-02654-25 Mfr # 840-298000
COMPARE
2 nm Dual beam 190
$7,194.00USD / Each
Thermo Scientific GENESYS™ 140 GENESYS™ 140 Visible Light Spectrophotometer, US plug EW-02654-26 Mfr # 840-308000
COMPARE
5 nm Dual beam 325
$5,835.00USD / Each
Thermo Scientific GENESYS™ 150 GENESYS™ 150 UV-Visible Light Spectrophotometer, US plug EW-02654-27 Mfr # 840-300000
COMPARE
2 nm Dual beam 190
$7,821.00USD / Each
Thermo Scientific GENESYS™ 180 GENESYS™ 180 UV-Visible Light Spectrophotometer, US plug EW-02654-28 Mfr # 840-309000
COMPARE
2 nm Dual beam 190
$8,827.00USD / Each
Thermo Scientific BioMate™ 160 BioMate™ 160 UV-Visible Light Spectrophotometer, US plug EW-02654-29 Mfr # 840-301000
COMPARE
2 nm Double beam 190
$8,391.00USD / Each
Showing 1- 7 of 7