Thermo Scientific Direct Heat Carbon Dioxide Incubators
High capacity chamber for maximum culture space
  • Lighter weight and more mobile than traditional water-jacketed designs
  • Intuative controls ensure easy operation
These incubators feature accurate control of temperature and CO2concentration. High performance heating elements and advanced insulation surrounding the culture chamber are excellent for temperature uniformity and recovery. Audible and visual alarms notify you of temperature deviations+/-1°C and CO2 deviations greater than 1% from set point, excluding deviations due to door openings. CO2 concentration ranges from 0 to 20% with +/-0.1% control. Units are stackable to conserve valuable lab space (hardware included).
Choose between the standard thermal conductivity (TC) sensor with accurate gas sensor technology, or the infrared (IR) sensor with exclusive split-beam technology – the perfect sensor for precise CO2 control in fluctuating temperature and humidity conditions.
$16,005.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (cu ft) Max Temperature (° C) Power (VAC)
Availability Pricing
COMPARE
6.5 50 115
$16,005.00USD / Each