This product is now discontinued, View Related Products
Thermo Scientific Compact Digital Rocking Shaker
Compact design easily fits in incubators (non-CO2), chambers, and refrigerators
  • PID control for a consistent smooth rocking motion
  • Digital speed control easily adjusts from 5 to 100 rpm
  • Adjustable tilt angle from 7 to 13° ensures gentle sample agitation
  • Use in timer mode up to 99 hrs 59 min or continuous rocking
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Load (lbs) Max Speed (rpm) Power (VAC)
Availability Pricing
Thermo Scientific 88880019 Compact Digital Rocking Shaker, 100-240V, US Plug. EW-04725-06 Mfr # 88880019
COMPARE
11 100 100 to 240
Call For Price
Thermo Scientific 88880020 Compact Digital Rocking Shaker, 100-240V, International Plug. EW-04725-07 Mfr # 88880020
COMPARE
11 100 100 to 240