This product is now discontinued, View Related Products
Thermo Scientific Cimarec Ultra-flat Stirrers with External Control
Powerful enough to stir large volumes and highly viscous fluids
  • Effectively stirs up to 10 liters
Stirrers feature a disc-drive motor that is both low-wear and powerful. The automatic start-up feautre allows for safe stirrer acceleration. There is no need to worry about breaking vessels with these units as the quick-stop function stops the drive within two seconds. External remote controller (sold separately) is connected via a 6-ft cable.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Stirring Volume (Liters)
Availability Pricing
Thermo Scientific Cimarec 50119111 Mobil 10 Large Volume Stirrer without controller EW-04651-85 Mfr # 50119111
COMPARE
10
Call For Price
Thermo Scientific Cimarec 50119112 Mobil 25 Large Volume Stirrer without controller EW-04651-86 Mfr # 50119112