Thermo Scientific Cimarec Single- and Multi-Position Stepper-Drive Stirrers with External Control
Heat — free operation preserves sensitive cultures
  • Made for long-term use in incubators
These Biosystem stirrers have a rugged hermatically sealed stainless steel housing that is impermeable to micro-organisms. The low-speed motor allows for even stirring to protect cells from damage.They require external remote controllers (sold separately).
External controllers are ideal for system integration. They have digital displays, high power settings for viscous media, and adjustable stirring and pause times with rotation sense inversion for synchronized operation of two stirring devices. They connect to the stirrer via a 6-ft cable. Choose from 115V or 230V with european plug.
$1,119.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
No. Of Stirring Positions
Availability Pricing
Thermo Scientific Cimarec 50119113 Biosystem Stirrer without controller EW-04651-83 Mfr # 50119113
COMPARE
1
$1,119.00USD / Each