Thermo Scientific Cimarec Mobil Direct Large Capacity Stirrers
Automatic start-up allows for safe stirrer acceleration
  • Low profile, compact design saves valuable bench space
These large-capacity stirrers feature a "quick-stop" function — stops stirring within three seconds, minimizing vessel breakage. Stainless steel housings is easy to clean and chemical resistant. Manufactured with a disc motor drive these stirrers are designed to be used with thick-bottomed vessels, round flasks, and double-walled containers — with stirring volumes of up to 150L. Integrated contoller has an adjustable power setting that decreases power consumption and lowers heat output.
$5,043.00 - $5,142.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC)
Availability Pricing
Thermo Scientific Cimarec 50088131 Mobil Direct 115 V US EW-04656-60 Mfr # 50088131
COMPARE
115
$5,142.00USD / Each
Thermo Scientific Cimarec 50088128 Mobile Direct Stirrer, 230V EW-04656-65 Mfr # 50088128
COMPARE
230
$5,043.00USD / Each