Thermo Scientific Cimarec Magnetic Microplate Shakers
Easily accommodates 96- or 384-well microplates
  • Durable aluminum nickel-coated housing is IP32 rated
Zero position, self-centering Thermo Scientific Cimarec® Magnetic Microplate Shakers have adjustable speed control. Automatic start-up enables slow start to prevent spilling of samples. Remote control units have an RS-232 interface — control up to 14 units with control unit or software (both included).
$3,635.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC)
Availability Pricing
Thermo Scientific 50094321-115V Teleshake, 100 to 2000 rpm, 115 VAC EW-51511-10 Mfr # 50094321
COMPARE
115
$3,635.00USD / Each