Thermo Scientific Barnstead Replacement Deionization Cartridges
Keep systems operating precisely
  • Ideal for high purity with lower capacity
  • Use for most analytical lab testing
These high capacity cartridge remove ionized impurities to produce high resistivity water with a neutral pH.
Model 01505-24 is ideal for high capacity, lower purity applications and compatible with the B-Pure, E-Pure, all Nanopure, and Bantam Barnstead systems.
Model 01506-26 and 01505-29 are suitable for high purity, lower capacity usage. The model 01506-26 cartridge is compatible with the B-Pure, Nanopure II, old Nanopure, and Bantam systems, while model 01505-29 is compatible with E-Pure and quad-style Nanopure.
$225.00 - $282.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Description Capacity (grains) Compatible with
Availability Pricing
Thermo Scientific Barnstead D0803 Deionization Cartridges, high capacity
Always in Stock
EW-01505-24 Mfr # D0803
COMPARE
Deionization Cartridges, high capacity 1700 as Calcium Carbonate All NANOpure™ E-pure™, B-pure™, Bantam systems
$265.00USD / Each
Thermo Scientific Barnstead D0809 Deionization Cartridge, high purity, 875 grain
Always in Stock
EW-01505-26 Mfr # D0809
COMPARE
Deionization Cartridge, high purity, 875 grain 875 as Calcium Carbonate NANOpure™ II, old NANOpure™, B-pure™
$282.00USD / Each
Thermo Scientific Barnstead D5027 Deionization Cartridge, high purity, 760 grain EW-01505-29 Mfr # D5027
COMPARE
Deionization Cartridge, high purity, 760 grain 760 as Sodium Chloride Quad-style NANOpure™, E-pure™
$225.00USD / Each