Thermo Scientific Barnstead Disposable Ion Exchange Cartridges
Customize your system with application-specific water purification cartridges
  • Disposable cartridges are ideal for use in nonpressurized applications
  • Inert polypropylene cartridge magerial prevents contamination
Order a cartridge holder (sold separately) to support your cartridges.
$140.00 - $446.00USD / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Description Capacity (grains) Media
Availability Pricing
Thermo Scientific Barnstead D8822 Ion Exchange Cartridge; Half-Size, Combo EW-99260-00 Mfr # D8822
COMPARE
Ion Exchange Cartridge; Half-Size, Combo 190 as Sodium Chloride Combination resin
$446.00USD / Each
Thermo Scientific Barnstead D50220 Ion Exchange Cartridge; Half-Size, Mixed-Bed EW-99260-01 Mfr # D50220
COMPARE
Ion Exchange Cartridge; Half-Size, Mixed-Bed 430 as Sodium Chloride Resin
$140.00USD / Each
Thermo Scientific Barnstead D8901 Ion Exchange Cartridge; Two-Bed EW-99260-02 Mfr # D8901
COMPARE
Ion Exchange Cartridge; Two-Bed 1725 as Sodium Chloride Resin
$237.00USD / Each
Thermo Scientific Barnstead D8950 Ion Exchange Cartridge; 2/3-Size, Two-Bed EW-99260-03 Mfr # D8950
COMPARE
Ion Exchange Cartridge; 2/3-Size, Two-Bed 1100 as Sodium Chloride Resin
$172.00USD / Each
Thermo Scientific Barnstead D8902 Ion Exchange Cartridge; Mixed-Bed EW-99260-04 Mfr # D8902
COMPARE
Ion Exchange Cartridge; Mixed-Bed 760 as Sodium Chloride Resin
$232.00USD / Each
Thermo Scientific Barnstead D8905 Disposable Ion Exchange Cartridge, cation removal EW-99260-06 Mfr # D8905
COMPARE
Disposable Ion Exchange Cartridge, cation removal 3000 as Sodium Chloride Resin
$159.00USD / Each
Thermo Scientific Barnstead D8904 Ion Exchange Cartridge, active charcoal EW-99260-08 Mfr # D8904
COMPARE
Ion Exchange Cartridge, active charcoal Activated charcoal
$199.00USD / Each
Thermo Scientific Barnstead D8903 Disposable Ion Exchange Cartridge, oxygen removal EW-99260-09 Mfr # D8903
COMPARE
Disposable Ion Exchange Cartridge, oxygen removal 30 g Resin
$246.00USD / Each
Thermo Scientific Barnstead D8911 Disposable Ion Exchange Cartridge, ultrapure EW-99260-10 Mfr # D8911
COMPARE
Disposable Ion Exchange Cartridge, ultrapure 915 as Sodium Chloride Resin
$293.00USD / Each
Thermo Scientific Barnstead D8921 Ion Exchange Cartridge, resin/active charcoal EW-99260-14 Mfr # D8921
COMPARE
Ion Exchange Cartridge, resin/active charcoal 1450 as Sodium Chloride Resin/activated charcoal
$205.00USD / Each
Thermo Scientific Barnstead D8922 Ion Exchange Cartridge; Mixed-Bed Combo EW-99260-16 Mfr # D8922
COMPARE
Ion Exchange Cartridge; Mixed-Bed Combo 600 as Sodium Chloride Resin/activated charcoal
$219.00USD / Each
Showing 1- 11 of 11