Testo Combustion Analyzers
Take hands-free readings - powerful magnets for mounting
  • Print measurement values wirelessly with the infrared output
  • Eliminate effect of excessive moisture on sensors with included condensate trap
  • Take meter anywhere - rechargeable batteries for easier portability
$905.00 - $1,115.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Display Type Includes Max Stack Temperature (° F)
Availability Pricing
Testo Model 310 Combustion Analyzer EW-86495-05 Mfr # 0563 3100 01
COMPARE
LCD Standard accessories 752
$905.00USD / Each
Testo Model 310 Combustion Analyzer Kit with infrared printer and two rolls of paper EW-86495-06 Mfr # 0563 3110 01
COMPARE
LCD Standard accessories plus infrared printer 752
$1,115.00USD / Each