Temperature Control Unit for Brookfield Rotational Viscometers
  • Measure hot melts, asphalt, wax, and polymers
This programmable unit elevates and controls the temperature of liquid samples. The solid-state proportional temperature controller and RTD probe maintain the stable temperature required for reproducible measuring of low-volume samples.
Use with small sample volumes fropm 8 to 13 mL. Viscosity range is 5 to 8,000,000 centipoise, depending on the spindle you install. Set one to ten temperature point programs. The thermo container reaches the set temperature in 30 minutes.
Order small sample adapters and spindles separately. Adapters attach to an external circulating water bath (not included) to cool the samples.
$6,628.00 - $7,745.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
Brookfield AMETEK HT-110-115 + HT-115A DP TEMP CONTROL W/SC4-18 SPNDL 115V/60HZ EW-98945-52 Mfr # SC4 -18 + HT-110-115 + HT-115
COMPARE
115
$7,745.00USD / Each
Brookfield AMETEK SC4-18 +HT-110-230 + HT-230A DP Temperature control unit; 230 VAC, 50 Hz EW-98945-54 Mfr # SC-18 + HT-110-230 + HT-230A DP
COMPARE
230
$6,628.00USD / Each