Jeio Tech Lab Companion Temperature and Environmental Chambers
Compact design conserves valuable laboratory space
  • Built-in battery and memory stores program patterns for 10 years without mains power
These temperature and environmental chambers are ideal when doing chemical experiments, animal and plant growth tests, microbial growth tests, and more. Insulated construction provides efficient heating and cooling with minimal temperature loss. Chambers feature a durable 304 stainless steel interior and painted steel exterior for harsh environments; standard 50-mm diameter cable port allows wiring to the interior. Tempered glass window provides a clear view of chamber during testing. Microprocessor
PID control with auto-tuning ensures stable control even in low temperature ranges. Enter and monitor the control parameters through the keypad and alphanumeric LCD. Input up to 30 programs with a total of 300 segments for all the programs.
$22,055.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Chamber Height (in) Chamber Width (in) Power (VAC)
Availability Pricing
Jeio Tech Lab Companion Temperature And Humidity Chamber 65 L, -20 C To 150°C; 220V 50/60 HZ EW-52401-94 Mfr # AAH821232K
COMPARE
21 19 3/4 220
$22,055.00USD / Each