This product is now discontinued, View Related Products
Techne Prime Thermal Cyclers
Choose from a reliable and robust selection of themal cycling options based on your needs
  • Colorful touch screen allows for quick set up and easy programming with intuitive software
  • Uses the Peltier effect — a chosen method for the most trusted thermal cyclers on the market
These high-performance thermal cyclers provide robust and reliable operation that is user friendly and easy on the pocketbook. Intuitive programming feature allows you to easily set up to 1000 programs with password protection to minimize inadvertent changes in settings. Use color touch screen to create complex programs. Heated lid has an adjustable temperature range of 35 to 115°C to prevent condensation in your samples. All units have the ability to transfer programs and temperature logging via the USB port between instruments or to your PC. An industry-leading 4-year warranty ensures years of reliable results.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Heating Rate Temperature Accuracy Description
Availability Pricing
COMPARE
up to 3.4⁰C per second ±0.2⁰C at 50⁰C Large-Format Thermal Cycler with Combi-Block; 100 to 230 V
Call For Price
COMPARE
up to 3.4⁰C per second ±0.2⁰C at 50⁰C Large-Format Gradient Thermal Cycler w/ Combi-Block; 100 to 230 V