Cole-Parmer® HI-200 Series Hybridization Incubators
Use and recover volumes as low as 5 mL, ensuring preservation of precious samples
  • Rotational speed from 0 to 20 rpm and rocking speed up to 60 opm
  • Radioactive-safe protective casing and nondrip tube design minimize risk to user
With excellent stability of ±0.1°C within the tubes, these hybridization incubators are great for nucleic hybridization applications such as Northern, Southern, and Western blots.
Each incubator features two shakers—rotisserie shaker with rotational speed from 0 to 20 rpm and platform shaker with rocking speed up to 60 oscillation/minute. A removable drip tray provides for easy cleaning. Adjustable feet make leveling on uneven surfaces quick and easy.
$4,135.00 - $5,930.00USD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Power (VAC) Description
Availability Pricing
COMPARE
100 120 Hybridization Incubator with Tubes, 100°C; 120 VAC
$5,620.00USD / Each
COMPARE
100 230 Hybridization Incubator, 100°C; 230 VAC
$5,020.00USD / Each
COMPARE
100 230 Hybridization Incubator with Tubes, 100°C; 230 VAC
$5,620.00USD / Each
COMPARE
80 230 Hybridization Incubator, 80°C; 230 VAC
$4,135.00USD / Each
COMPARE
80 230 Hybridization Incubator with Tubes, 80°C; 230 VAC
$4,925.00USD / Each
COMPARE
80 120 Hybridization Incubator, 80°C; 120 VAC
$4,190.00USD / Each
COMPARE
80 120 Hybridization Incubator with Tubes, 80°C; 120 VAC
$5,930.00USD / Each
Showing 1- 7 of 7