Cole-Parmer® Syringe Filters, Non-Sterile
Economical, single use, and efficient filtering stream for high output filtration from viscous and particle-laden solutions
  • Reduce bursting with reinforced polypropylene housing Take advantage of a large effective filtration area Standard female luer-lock inlet and male luer-slip outlet Available in various membrane size, pore size, and material combinations
Use the single-use syringe filters for clarification and prefiltration, and even for removing fine particulate with the membrane’s large effective filtration area. Reinforced polypropylene housing has secure connections for male luer slip outlet and female luer lock inlet. Non-sterile bulk pack of syringe filters 100 per pack. The membrane type and pore size are visible on each filter housing as well.
$129.25 - $236.25USD / Pkg of 100
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Membrane Diameter (mm) Membrane Material Pore Size (µm)
Availability Pricing
COMPARE
25 PES 0.2
$129.25USD / Pkg of 100
COMPARE
25 PES 0.45
$129.25USD / Pkg of 100
COMPARE
25 PES with prefilter 0.2
$164.75USD / Pkg of 100
COMPARE
25 PES with prefilter 0.45
$164.75USD / Pkg of 100
COMPARE
25 glass fiber 1.5
$236.25USD / Pkg of 200