This product is now discontinued, View Related Products
Swift Optical Compound Microscope with Tablet Display and Camera
Includes everything you need to view and capture images
  • Edit images directly on the tablet touchscreen
  • View images comfortably on large, 8" display
  • Share images with others using built-in Wi-Fi
These microscopes are an all-in-one image capture solution - built-in 8" (20.3 cm) LCD display allows you to view your specimen while the internal camera captures the image. Edit, measure, and annotate saved images directly on the touchscreen or on a computer with the included software. Display features an Android operating system and a micro-SD card slot for additional memory (4 GB SD card included). Wi-Fi signal connects to a computer for image download and sharing. Siedentopf binocular head allows for easy adjustability and the 20-mm eyepieces provide an increased field of view.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Objectives
Availability Pricing
Swift Optical Compound microscope with Tablet-Style Display and Camera, Semi-plan objectives EW-48925-11 Mfr # M10T-BTW1-S
COMPARE
Semi-plan
Call For Price
Swift Optical Compound Microscope with Tablet-Style Display and Camera, Plan objectives EW-48925-12 Mfr # M17T-BTW-P
COMPARE
Plan
Call For Price
Swift Optical Compound Microscope with Tablet-Style Display and Camera, Phase-Contrast objectives EW-48925-13 Mfr # M10T-BTW1-MP
COMPARE
Phase contrast