Sussman Low-Capacity Electric Steam Generators
Low-cost steam source for heating, cleaning, and sterilizing
  • Convert approximately 97% of electric energy to steam energy
Use these steam generators in industries that require efficient, high-quality steam. Perfect for laboratories, computer rooms, and pilot plants; and for small, intermittent, low-steam consuming loads. Generators supply a reliable, constant steam source for use with your steam tables, distillation apparatus, autoclaves, and other equipment.
Produce full pressure in minutes without the fumes and combustion compounds produced by fossil fuel powered generators. Cool, silent, odorless operation is virtually effortless—connect the electrical and water supplies and set the pressure control valve. Minimal maintenance and simple blowdown procedure ensure years of trouble-free operation.
Liquid-level controller automatically maintains proper water level and shuts off generator if water supply drops below safe level; pressure control feature provides high-limit pressure control with manual reset. All models are built to ASME Section I, Code M standards and are National Board registered. Electrical connections conform to NEC standards.
Generators come equipped with steam pressure gauge, water strainer, water level sight glass, and an on/off pilot light. Steam outlet is 1/2" NPT(F); water connection is 1/4" NPT(F). Generators require an inlet water pressure of 10 psi greater than the desired maximum steam outlet pressure. If operating pressure exceeds incoming water pressure, a motor and pump are required; see our "Pumps" section.
$5,980.00 - $6,815.00USD / Each
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Electric Capacity (kW) Output (lbs/hr) Power (VAC)
Availability Pricing
Sussman MBA9B3 Electric Steam Generator, with 120 VAC control circuit, 27.0 lb/hr, 208 VAC EW-07602-00 Mfr # MBA9B3
COMPARE
9 27 208
$6,210.00USD / Each
Sussman MBA9C3 Electric Steam Generator, with 120 VAC control circuit, 27.0 lb/hr, 240 VAC EW-07602-10 Mfr # MBA9C3
COMPARE
9 27 240
$6,205.00USD / Each
Sussman MBA12B3 Electric Steam Generator, with 120 VAC control circuit, 36.0 lb/hr, 208 VAC EW-07602-20 Mfr # MBA12B3
COMPARE
12 36 208
$6,095.00USD / Each
Sussman Electric Steam Generator, with 120 VAC control circuit, 36.0 lb/hr, 240 VAC EW-07602-30 Mfr # MBA12C3 / 3 PHASE
COMPARE
12 36 240
$5,980.00USD / Each
Sussman MBA3A1 Electric Steam Generator, with 120 VAC control circuit, 9.0 lb/hr, 120 VAC EW-07603-00 Mfr # MBA3A1
COMPARE
3 9 120
$6,000.00USD / Each
Sussman MBA6B3 Electric Steam Generator, with 120 VAC control circuit, 18.0 lb/hr, 208 VAC EW-07603-10 Mfr # MBA6B3
COMPARE
6 18 208
$6,440.00USD / Each
Sussman MBA6C3 Electric Steam Generator, with 120 VAC control circuit, 18.0 lb/hr, 240 VAC EW-07603-20 Mfr # MBA6C3
COMPARE
6 18 240
$6,395.00USD / Each
Sussman MBA20F3 Electric Steam Generator, with 120 VAC control circuit, 60.0 lb/hr, 480 VAC EW-07603-40 Mfr # MBA20F3
COMPARE
20 60 480
$6,540.00USD / Each
Sussman MBA18C3 Electric Steam Generator, with 120 VAC control circuit, 54.0 lb/hr, 240 VAC EW-07603-50 Mfr # MBA18C3
COMPARE
18 54 240
$6,745.00USD / Each
Sussman MBA18F3 Electric Steam Generator, with 120 VAC control circuit, 54.0 lb/hr, 480 VAC EW-07603-52 Mfr # MBA18F3
COMPARE
18 54 480
$6,815.00USD / Each
Sussman MBA3C1 Electric Steam Generator, with 120 VAC control circuit, 9.0 lb/hr, 240 VAC EW-07604-00 Mfr # MBA3C1
COMPARE
3 9 240
$6,240.00USD / Each
Sussman MBA6C3EX Electric Steam Generator, with 240 VAC control circuit, 18.0 lb/hr, 240 VAC EW-07604-15 Mfr # MBA6C3EX
COMPARE
6 18 240
$6,395.00USD / Each
Sussman MBA9C3/EX Electric Steam Generator, with 240 VAC control circuit, 27.0 lb/hr, 240 VAC EW-07604-20 Mfr # MBA9C3/EX
COMPARE
9 27 240
$6,165.00USD / Each
Sussman MBA12C3/EX Electric Steam Generator, with 240 VAC control circuit, 36.0 lb/hr, 240 VAC EW-07604-30 Mfr # MBA12C3/EX
COMPARE
12 36 240
$6,000.00USD / Each
Showing 1- 14 of 14