Submersible Pumps for Corrosive/Acid Sumps
Only electric submersible pump with entire motor encapsulated in Noryl®
  • Noryl material never rusts and provides excellent resistance against acids and alkalis
  • Compact and lightweight—only 13-1⁄2 lbs—with convenient carrying handle
  • Virtually unbreakable
These pumps can easily be installed in a sump and are ideal for safeguarding vaults, emptying tanks, or pumping out ponds. The unique dual-port design virtually eliminates kinks in your hose; use vertical port for deep/narrow places or use the horizontal port for shallow/wide applications. Thermal breaker incorporated in motor windings shuts off power before over-heating occurs due to locked rotor or long periods of running dry.
For permanent installation, temperature should not exceed 135°F (57°C). For intermittent use, submerge less than 20 minutes in 195°F (90°C) maximum.
$1,182.50 - $2,463.50USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (GPM) Power (VAC) Motor hp
Availability Pricing
COMPARE
47 115 0.4
$1,346.00USD / Each
COMPARE
47 115 0.4
$1,182.50USD / Each
COMPARE
47 115 0.4
$1,394.50USD / Each
COMPARE
47 115 0.4
$1,346.50USD / Each
COMPARE
47 115 0.4
$2,463.50USD / Each