Cole-Parmer® SH-200 Series See-Saw Rocking Shakers
Gently mix live cultures
  • Nonslip mat and platform lip hold vessels securely in place
  • Built-in timer and digital speed LED display provide reproducible results
  • Can be used in incubators
These see-saw rocking shakers provide a gentle rocking action at a 7° tilt angle, perfect for culture flasks and Petri dishes. Easily set and view the speed and the timer on the LED display. Control speed from 5 to 70 rpm. The microprocessor maintains speed accuracy even over long runs. Easily set digital timer from 1 to 999 minutes. When timer reaches zero, the shaker automatically stops and sounds an alert.
Durable shakers are rated for continuous use. Use in incubators and environmental or cold chambers up to 104°F (40°C).
$1,052.00 - $1,396.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Load (lbs) Platform Width (in) Power (VAC)
Availability Pricing
COMPARE
22 14 230
$1,396.00USD / Each
COMPARE
22 14 120
$1,213.00USD / Each
COMPARE
6.6 9.25 230
$1,071.00USD / Each
COMPARE
6.6 9.25 120
$1,052.00USD / Each