Cole-Parmer® Heating Blocks for Hot Plates and Stirring Hot Plates
Turn your existing 6" x 6" hot plate into a heating mantle
  • Safest way to heat round-bottom flasks
  • Flexible modular system for a wide range of sizes up to 500 mL
  • Low-well design is perfect for multi-neck flasks
Designed to heat round-bottom flasks, the heating block system with interchangeable inserts can be used with various Cole-Parmer hot plates and hot plate stirrers with 6" x 6" top plate. The heating block system offers two sizes of base plates (small and medium) which can accommodate up to three block inserts each. A third larger base plate features heat-resistant handles and accepts a single block insert for flasks up to 500 mL. Chemical- and solvent-resistant anodized block inserts provide a safe method for synthesis of single or multiple reactions and offer equal or better heating performance than silicon oil. The low-well provides excellent temperature transfer without compromising the view of the reaction or risk of cracking the glassware. Each base plate features a hole to house a temperature controller probe.
$225.00 - $533.00USD / Each
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Heater Block Base for Hot Plate or Stirring Hot Plate, Large
$533.00USD / Each
COMPARE
Heater Block Base for Hot Plate, Small
$350.00USD / Each
COMPARE
Heater Block Insert for Large Base, 100 mL, 1/EA
$232.00USD / Each
COMPARE
Heater Block Clamp for Small Base
$276.00USD / Each
COMPARE
Heater Block Insert for Small & Medium Bases, 100 mL, 3/PK
$431.00USD / Each
COMPARE
Heater Block Insert for Small & Medium Bases, 10 mL, 3/PK
$375.00USD / Each
COMPARE
Heater Block Base for Hot Plate or Stirring Hot Plate, Medium
$449.00USD / Each
COMPARE
Heater Block Insert for Large Base, 250 mL, 1/EA
$232.00USD / Each
COMPARE
Heater Block Clamp for Medium Base
$258.00USD / Each
COMPARE
Heater Block Insert for Medium Base, 250 mL, 3/PK
$524.00USD / Each
COMPARE
Heater Block Insert for Small & Medium Base, 25 mL, 3/PK
$375.00USD / Each
COMPARE
Heater Block Insert for Large Base, 500 mL, 1/EA
$225.00USD / Each
COMPARE
Heater Block Insert for Large Base, 50 mL, 1/EA
$232.00USD / Each
COMPARE
Heater Block Insert for Small & Medium Bases, 50 mL, 3/PK
$392.00USD / Each
COMPARE
Heater Block Insert for Small & Medium Bases, 5 mL, 3/PK
$369.00USD / Each
Showing 1- 15 of 15