This product is now discontinued, View Related Products
Stuart Antiglare Ultraviolet Transilluminators
Exceptional performance in a compact size
  • Antiglare surface ensures uniform UV light distribution
  • Flexible—switch from high (100%) to low (50%) intensity
  • Lamps arranged in an array of 6 x 8 W for 48 W total output
The compact size of the transilluminator makes it easy to maneuver, while at the same time saving valuable benchtop space. Designed specifically for DNA and RNA work, the units feature a dual-intensity control switch to choose either a high or low intensity and optimized antiglare filter glass area of 20 x 24 cm that provides uniform illuminations.
Call For Price
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Wavelength Description
Availability Pricing
COMPARE
110 254 nm UV Transilluminator, 6 x 8 W Lamps, Antiglare, 254 nm, 20 x 24 cm Filter; 110 VAC, 60 Hz
Call For Price
COMPARE
110 312 nm UV Transilluminator, 6 x 8 W Lamps, Antiglare, 312 nm, 20 x 24 cm Filter; 110 VAC, 60 Hz
Call For Price
COMPARE
110 364 nm UV Transilluminator, 6 x 8 W Lamps, Antiglare, 365 nm, 20 x 24 cm Filter; 110 VAC, 60 Hz
Call For Price
COMPARE
220 254 nm UV Transilluminator, 6 x 8 W Lamps, Antiglare, 254 nm, 20 x 24 cm Filter; 220 VAC, 50 Hz Euro
Call For Price
COMPARE
220 312 nm UV Transilluminator, 6 x 8 W Lamps, Antiglare, 312 nm, 20 x 24 cm Filter; 220 VAC, 50 Hz Euro
Call For Price
COMPARE
220 365 nm UV Transilluminator, 6 x 8 W Lamps, Antiglare, 365 nm, 20 x 24 cm Filter; 220 VAC, 50 Hz Euro
Call For Price
COMPARE
220 254 nm UV Transilluminator, 6 x 8 W Lamps, Antiglare, 254 nm, 20 x 24 cm Filter; 220 VAC, 50 Hz UK
Call For Price
COMPARE
220 312 nm UV Transilluminator, 6 x 8 W Lamps, Antiglare, 312 nm, 20 x 24 cm Filter; 220 VAC, 50 Hz UK
Call For Price
COMPARE
220 365 nm UV Transilluminator, 6 x 8 W Lamps, Antiglare, 365 nm, 20 x 24 cm Filter; 220 VAC, 50 Hz UK
Call For Price
COMPARE
220 254 nm UV Transilluminator, 6 x 8 W Lamps, Antiglare, 254 nm, 20 x 24 cm Filter; 220 VAC, 50 Hz SA
Call For Price
COMPARE
220 312 nm UV Transilluminator, 6 x 8 W Lamps, Antiglare, 312 nm, 20 x 24 cm Filter; 220 VAC, 50 Hz SA
Call For Price
COMPARE
220 365 nm UV Transilluminator, 6 x 8 W Lamps, Antiglare, 365 nm, 20 x 24 cm Filter; 220 VAC, 50 Hz SA
Call For Price
Showing 1- 12 of 12