Stopcocks with Flow Indicators
PVC bodies, EPDM seals and seats. Flow indicator on handle. Maximum pressure is 150 psi at the maximum temperature of 120°>F (49°>C); maximum vacuum is 29.9'' Hg.
$34.50 - $42.00USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connection Type Orifice (in)
Availability Pricing
COMPARE
NPT(F) 0.25
$42.00USD / Each
COMPARE
NPT(M) 0.25
$34.50USD / Each
COMPARE
NPT(M) x Barbed 0.25
$42.00USD / Each
COMPARE
Barbed 0.25 to 0.5
$41.50USD / Each
COMPARE
NPT(M) x NPT(F) 0.25
$42.00USD / Each