Stopcocks with Compression Fittings 48575
Stopcocks have body made of PFA and plug made of PTFE. Maximum pressure is 50 psi.
$127.00 - $130.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Cv Tubing OD (in)
Availability Pricing
COMPARE
0.38 0.125
$130.00USD / Each
COMPARE
0.44 0.25
$127.00USD / Each