Swift Optical SM100 Series LED Stereo and Stereozoom Microscopes
Features top, bottom, and turret lights to meet all your illumination needs
  • Comfortable 45° inclined tubes with 360° rotatable head minimizes strain
  • Energy-efficient variable LED illumination with 5 light setting combinations provides bright, white light
  • Cordless microscope uses rechargeable batteries for easy field use
  • Built-in handle encourages proper handling and makes transportation easy
Swift Optical SM100 Series microscopes feature variable LED illumination and cordless operation for added versatility. Energy-efficient LEDs provide bright white light with virtually no heat — ideal for temperature-sensitive samples. Five light setting combinations with top, bottom, and turret lights and dimmer control allow for varying light intensity. Microscope head features interlocked eyepiece tubes, 10X/20 mm eyepieces, and diopter adjustment. Magnification control is mounted on the side of the head for easy adjustments.
Frosted glass and black and white stage plates are provided with the microscope stand. The frosted glass stage plate allows light from the bottom illuminator to pass through. The black and white stage plates can be used for additional contrast to the specimen that is being viewed.
$422.00 - $696.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Head Configuration Magnification Objectives
Availability Pricing
Swift Optical SM101-C Stero Microscope, LED, Binocular, 10X and 30X; Rechargeable EW-03904-23 Mfr # SM101-C
COMPARE
Binocular 10X and 30X 1X and 3X
$422.00USD / Each
Swift Optical SM105-C Stereozoom Microscope, LED, Binocular, 10X to 30X Zoom; Rechargeable EW-03904-25 Mfr # SM105-C
COMPARE
Binocular 10X to 30X Zoom 1X, 2X, and 3X Zoom
$593.00USD / Each
Swift Optical SM105T-C Stereozoom Microscope, LED, Trinocular, 10X to 30X Zoom; Rechargeable EW-48926-71 Mfr # SM105T-C
COMPARE
Trinocular 10X to 30X Zoom 1X, 2X, and 3X Zoom
$696.00USD / Each