This product is now discontinued, View Related Products
Starrett Professional Dial Calipers
Versatility and accuracy in a compact design
  • Sharp black graduations on the satin finished bar for quick measurement readings
  • Positive, spring-loaded, double pinion, anti-backlash control
This dial caliper is one of the handiest measuring tools available. Used by mechanics and tool-makers everywhere, this tool has knife edge contacts for inside and outside measurements. It is useful for scribing lines parallel to work edges. Hardened stainless steel rack guarantees lasting accuracy.
The calipers feature knife-edge contacts, thumb-operated fine adjustment roll, adjustable bezel and lock screws for sliding jaw and dial bezel.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max range (in) Description
Availability Pricing
Starrett 120A-6 Dial Calipers Stainless Steel 0-6" EW-09925-06 Mfr # 120A-6
COMPARE
6 Dial Calipers Stainless Steel 0-6\"
Call For Price
Starrett 120A-9 Dial Calipers, Stainless Steel 0-9" EW-09925-07 Mfr # 120A-9
COMPARE
9 Dial Calipers, Stainless Steel 0-9\"
Call For Price
Starrett 120-12 Dial Calipers, Stainless Steel, 0-12" EW-09925-08 Mfr # 120-12
COMPARE
12 Dial Calipers, Stainless Steel, 0-12\"