Standard RC Dialysis Trial Kits
Contains everything you needed to evaluate dialysis membranes
  • Ideal for smaller dialysis experiments
These convenient kits contain everything you need for smaller dialysis experiments or evaluating dialysis tubing for a dialysis application. The small roll is ideal for low volume projects. Spectra/Por® 1 through 3 Dialysis Trail Kits are available in a range of nominal molecular weight cut-offs (MWCO) for less specific separations. Spectra/Por grades 1 through 3 are packaged dry with 10% glycerol and contain trace levels of heavy metals and sulfur.
$180.00 - $195.00USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Flat Width (mm) Max Molecular Weight Cut Off (Mwco) (Dalton) Min Molecular Weight Cut Off (Mwco) (Dalton)
Availability Pricing
Spectra Por S/P 1 Dialysis Membrane Trial Kit, 6000-8000 Dalton, 32mm Flat Width EW-02900-00 Mfr # 132655T
COMPARE
32 8000 6000
$180.00USD / Each
Spectra Por S/P 1 Dialysis Membrane Trial Kit, 6000-8000 Dalton, 50mm Flat Width EW-02900-01 Mfr # 132665T
COMPARE
50 8000 6000
$195.00USD / Each
Spectra Por S/P 2 Dialysis Membrane Trial Kit, 12,000-14,000 Dalton, 25mm Flat Width EW-02900-02 Mfr # 132678T
COMPARE
25 14000 12000
$180.00USD / Each
Spectra Por 2 Dialysis Membrane Trial Kit, 12,000-14,000 Dalton, 50mm Flat Width EW-02900-03 Mfr # 132680T
COMPARE
45 14000 12000
$195.00USD / Each
Spectra Por S/P 3 Dialysis Membrane Trial Kit, 3,500 Dalton, 18mm Flat Width EW-02900-04 Mfr # 132720T
COMPARE
18 3500 3500
$180.00USD / Each
Spectra Por S/P 3 Dialysis Membrane Trial Kit, 3,500 Dalton, 54mm Flat Width EW-02900-05 Mfr # 132725T
COMPARE
54 3500 3500
$195.00USD / Each