Standard Dial Durometers
Meet your specific application
  • Compliance: ASTM D-2243
Standard Dial Durometers feature a large, easy-to-read dial, excellent 1/2 point accuracy, and full 360° sweep. Scale is divided into 1-point increments from 0 to 100. Measures 2-1/2"W x 6-1/8"H.
$595.50 - $1,141.00USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Calibration Included Type
Availability Pricing
Rex Gauge 1600 TYPE A Standard Dial Hardness Tester/Durometer; Type A EW-59792-08 Mfr # 1600 TYPE A
COMPARE
No A
$624.00USD / Each
Rex Gauge 1600 TYPE D Standard Dial Hardness Tester/Durometer; Type D EW-59792-10 Mfr # 1600 TYPE D
COMPARE
No D
$595.50USD / Each
Rex Gauge MICRO A 1600 Standard Dial Hardness Tester/Durometer; Type M (Micro-A) EW-59792-16 Mfr # MICRO A 1600
COMPARE
No M
$1,141.00USD / Each
Rex Gauge 1600 TYPE A Standard Dial Durometer with NIST Calibration Certificate, Type A EW-59792-36 Mfr # 1600 TYPE A W/NIST CERT
COMPARE
Yes A
$616.50USD / Each
Rex Gauge 1600 TYPE D Standard Dial Durometer with NIST Calibration Certificate, Type D EW-59792-38 Mfr # 1600 TYPE D W/NIST CERT
COMPARE
Yes D
$605.50USD / Each
Rex Gauge MICRO A 1600 Standard Dial Durometer with NIST Calibration Certificate, Micro A EW-59792-44 Mfr # MICRO A 1600 W/NIST CERT
COMPARE
Yes M
$1,077.00USD / Each