Spilhyder Universal SM/SMS Sorbent Rolls
Coverstock uses spunbond material for excellent durability and virtually no linting
Universal sorbents are ideal for environments where there are many different liquids including cutting oils, lubricants, and coolants. Works with both water-based liquids, oil-based liquids, and more. Spunbond coverstock is abrasion resistant - withstands rigorous abuse from heavy foot traffic. High strength will resist tearing when placed under equipment.
$73.00 - $273.75USD / Bag of 1
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Absorption Description
Availability Pricing
NPS Spilfyter Spilhyder Universal Gray Spill Pad Roll, Medium-Weight, 32" x 150', 1/Bag EW-86423-63 Mfr # SM-90
COMPARE
56 gal. (212 L) Universal Gray Spill Pad Roll, Medium-Weight, 32\" x 150', 1/Bag
$273.75USD / Bag of 1
NPS Spilfyter Spilhyder All-in-One Spill Pad Roll, Medium-Weight, 15" x 75', 1/Case EW-86423-66 Mfr # DB-1575
COMPARE
10 gal. (38 L) All-in-One Spill Pad Roll, Medium-Weight, 15\" x 75', 1/Case
$73.00USD / Case of 1
NPS Spilfyter Spilhyder All-in-One Spill Pad Roll, Medium-Weight, 16" x 150', 1/Case EW-86423-67 Mfr # DB-914
COMPARE
28 gal. (106 L) All-in-One Spill Pad Roll, Medium-Weight, 16\" x 150', 1/Case
$150.50USD / Case of 1