Spilfyter Test Strips and Reference Charts
Classify unknown spills in 30 seconds
Convenient strips change color to identify risk type and classify spill for proper treatment. Compare color indicator to laminated, reusable reference chart to classify unknown liquid. Chemical classifier kit tests for acid or base, iodine, oxidizers, chlorine, fluoride, bromine and petroleum/-organic products. Wastewater classifier kit tests for acid/base, petroleum/organic solvent, hydrogen sulfide, nitrite, nitrate, and fluoride risks.
$9.90 - $808.50USD / Case of 50
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
NPS Spilfyter Chemical test strips, case of 50 EW-86423-50 Mfr # 570050
COMPARE
Chemical test strips, case of 50
$808.50USD / Case of 50
NPS Spilfyter Chemical Reference Chart EW-86423-54 Mfr # 577777
COMPARE
Chemical Reference Chart
$9.90USD / Each
NPS Spilfyter Wastewater Reference Chart EW-86423-56 Mfr # 588888
COMPARE
Wastewater Reference Chart
$27.80USD / Each
NPS Spilfyter Chemical Classifier Strips, 10/Pack EW-86424-15 Mfr # 570010
COMPARE
Chemical Classifier Strips, 10/Pack
$144.50USD / Pkg of 10
NPS Spilfyter Wastewater Classifier Strips, 10/Pack EW-86424-17 Mfr # 580010
COMPARE
Wastewater Classifier Strips, 10/Pack
$159.75USD / Pkg of 10