Spilfyter Kolor-Safe Acid Neutralizers
Three-color change lets you know when spill is completely neutralized
Protect your workers and facility from acid spills by quickly and efficiently neutralizing them. Simply apply the neutralizer until the spill goes through the three-color change. Acid neutralizer changes from purple to yellow to a reddish color to indicate total neutralization.
$32.60 - $71.40USD / Gallon of 1
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
NPS Spilfyter Kolor-Safe Acid Neutralizer, 1 quart EW-86423-10 Mfr # 410001
COMPARE
Acid Neutralizer, 1 quart
$32.60USD / Quart of 1
NPS Spilfyter Kolor-Safe Acid Neutralizer, 1 gallon EW-86423-11 Mfr # 410004
COMPARE
Acid Neutralizer, 1 gallon
$71.40USD / Gallon of 1