Cole-Parmer® VapLock™ Solvent Waste Kits for Jars and Pails
Collect effluent from multiple sources while minimizing harmful solvent exhaust
  • Various cap sizes available to fit your solvent container
  • 10 port manifold—1 x 1/2" NPT(F), 3 x 1/4" NPT(F), and 6 x 1/4-28 UNF(F)
Use the waste kits to minimize leaks, spills, and vapor escape from solvent waste containers. Waste kits include a manifold, exhaust filter, adapters, plugs, and tubing—everything you need to install on a typical HPLC instrument. Manifold allows solvent waste to be collected from multiple effluent streams. Exhaust filter minimize exposure to harmful chemical vapors. Fittings and tubing provide leak-tight connections minimizing spills and leaks.
$378.25 - $563.00USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cap / Stopper Size Material
Availability Pricing
COMPARE
70 mm Rieke 8 TPI Polypropylene(PP)
$408.75USD / Each
COMPARE
70 mm Rieke 8TPI Polytetrafluoroethylene (PTFE)
$453.25USD / Each
COMPARE
100-415 Polypropylene(PP)
$407.50USD / Each
COMPARE
100-415 Polytetrafluoroethylene (PTFE)
$428.50USD / Each
COMPARE
70 mm Rieke 6TPI Polypropylene(PP)
$378.25USD / Each
COMPARE
70 mm Rieke 6TPI Polytetrafluoroethylene (PTFE)
$563.00USD / Each