Cole-Parmer® Spex® Grinding Vial Sets
Ideal for pulverizing the toughest rocks, mineral, cement, and ceramics
  • Help reduce samples to analytical powders for XRF and ICP
  • Use with vial holder 04575-32
Each vial measures 2-1/8"L x 3/4" dia. Grinding load is 0.5 to 1.5 g.
$6.70 - $530.50USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description Special Features
Availability Pricing
COMPARE
Stainless Steel Capped Vial Set for the GenoLyte or MixerMill Caps with stainless steel inserts
$366.75USD / Each
COMPARE
Tungsten Carbide-Lined Vial Set for the GenoLyte or MixerMill Caps with tungsten carbide inserts
$530.50USD / Each
COMPARE
Stainless Steel Grinding Ball, 5/16\" (7.9 mm) Stainless steel grinding ball
$6.70USD / Each
COMPARE
Tungsten Carbide Grinding Ball, 5/16\" (7.9 mm) Tungsten carbide grinding ball
$37.60USD / Each