Spex CertiPrep® Assurance® Grade Quality Control Standard 7A
Multi-element standards for AA & ICP instrumentation
  • Quality control standards help ensure accurate and reliable results
  • Manufactured under ISO 9001, ISO/IEC 17025 and ISO 17034 compliance
Use the Quality Control Standards to check the standard curve, the procedure for inter-element correction and other spectral interferences. These standards are carried through the entire analytical operation of the method. If the determined concentration is not within ± 5% of 1 µg/mL, the laboratory performance is unacceptable. The source of the problem should be identified and corrected before continuing the analysis.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
$226.00 - $379.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Concentration Volume (mL) Description
Availability Pricing
COMPARE
Multiple 125 Assurance® Grade Quality Control Standard 7A for AA & ICP in 5% HNO3/tr. HF; 125 mL
$226.00USD / Each
COMPARE
Multiple 500 Assurance® Grade Quality Control Standard 7A for AA & ICP in 5% HNO3/tr. HF; 500 mL
$379.00USD / Each