Speirs Robertson AMD-SM Series Microscope Isolation Tables
Create the best working environment for your microscope
  • Range of microscope tables incorporating high performance vibration isolation
  • Custom designed elastomeric isolators built into worktop
  • Ergonomic design for best user comfort, maximising accuracy and efficiency
Get the best performance out of your microscope by placing in an environment where performance is not compromised removing unwanted vibrations. We offer a choice of isolators to provide different levels of isolation from that required for laboratory microscopes up to the highest performance instruments such laser confocal and spinning disk microscopes. Range of sizes and accessories allows you to make a custom table that suits your requirements. Front guards are included and protect against accidental knocks. Tables are simple to install, easy to use and maintenance free.
$2,047.94 - $2,089.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Width (in) Capacity (kg) Height (in)
Availability Pricing
Speirs Robertson AMD-SM-75x75 AMD-SM Microscope Table, Sorbothane Isolators; 75x75 cm EW-11500-29 Mfr # AMD-SM-75X75
COMPARE
30 75 30
$2,089.00USD / Each
Speirs Robertson AMD-SM-90x75 AMD-SM Microscope Table, Sorbothane Isolators; 90x75 cm EW-11500-31 Mfr # AMD-SM-90X75
COMPARE
36 75 30
$2,080.23USD / Each
Speirs Robertson AMD-SM-120x75 AMD-SM Microscope Table, Sorbothane Isolators; 120x75 cm EW-11500-32 Mfr # AMD-SM-120X75
COMPARE
48 75 30
$2,047.94USD / Each