Speirs Robertson ALSC Series Draft Shield / Acoustic Hoods
Maximise instrument performance by removing airborne vibrations
  • High quality range of hoods made from interlocking clear acrylic panels
  • Excellent all round visibility prevents errors
  • Lift off front panel with built in access ports
  • Pharma stainless base tray included
Acoustic vibrations from fans, doors and other sources can prevent accurate measurements by disturbing equipment and causing temperature changes. The acrylic hoods are simply placed over the equipment onto a worktop and prevent disturbances reaching sensitive instruments such as balances and microscopes. Hoods should be placed on a flat surface using the base tray provided.
$1,677.54 - $2,096.92USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Draft Shield Height (in) Draft Shield Width (in) Width (in)
Availability Pricing
Speirs Robertson ALSC-75x75 ALSC Clear Acrylic Draft Shield / Acoustic Hood75x60x75 cm EW-11500-64 Mfr # ALSC-75X75
COMPARE
23 5/8 29 33/64 30
$1,677.54USD / Each
Speirs Robertson ALSC-90x75 ALSC Clear Acrylic Draft Shield / Acoustic Hood90x60x75 cm EW-11500-65 Mfr # ALSC-90X75
COMPARE
23 5/8 35 27/64 36
$1,770.74USD / Each
Speirs Robertson ALSC-120x75 ALSC Clear Acrylic Draft Shield / Acoustic Hood120x60x75 cm EW-11500-66 Mfr # ALSC-120X75
COMPARE
23 5/8 47 1/4 48
$2,096.92USD / Each