Spectra/Por 7 Pretreated Regenerated Cellulose (RC) Tubing
Available in a variety of specific MWCO's
  • Ideal for separations sensitive to heavy metal traces
For separations that require specific retentivity and are sensitive to sulfur and metals. EDTA-treated membranes contain no heavy metal traces or sulfur. Supplied wet in 0.1% sodium azide solution. Ready for use after rinsing and conditioning in dialyzing buffer. Each box contains 16-ft of membrane.
$332.00 - $411.00USD / Box of 1
22 variations of this product are available.
See All Product Options Below
22 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Flat Width (mm) Max Molecular Weight Cut Off (Mwco) (Dalton)
Availability Pricing
Spectra Por S/P 7 7 RC Dialysis Tubing, 2000 MWCO, 18mm/11.5mm; 16ft EW-02890-43 Mfr # 132107
COMPARE
11.5 18 2000
$411.00USD / Box of 1
Spectra Por S/P 7 7 RC Dialysis Tubing, 2000 MWCO, 38mm/24mm; 16ft EW-02890-44 Mfr # 132108
COMPARE
24 38 2000
$411.00USD / Box of 1
Spectra Por S/P 7 7 RC Dialysis Tubing, 2000 MWCO, 45mm/29mm; 16ft EW-02890-46 Mfr # 132109
COMPARE
29 45 2000
$411.00USD / Box of 1
Spectra Por S/P 7 7 RC Dialysis Tubing, 3500 MWCO, 18mm/11.5mm; 16ft EW-02890-47 Mfr # 132110
COMPARE
11.5 18 3500
$374.00USD / Box of 1
Spectra Por S/P 7 7 RC Dialysis Tubing, 3500 MWCO, 45mm/29mm; 16ft EW-02890-48 Mfr # 132111
COMPARE
29 45 3500
$411.00USD / Box of 1
Spectra Por S/P 7 7 RC Dialysis Tubing, 3500 MWCO, 54mm/34mm; 16ft EW-02890-49 Mfr # 132112
COMPARE
34 54 3500
$411.00USD / Box of 1
Spectra Por S/P 7 7 RC Dialysis Tubing, 8000 MWCO, 24mm/15mm; 16ft EW-02890-51 Mfr # 132114
COMPARE
15 24 8000
$332.00USD / Box of 1
Spectra Por S/P 7 7 RC Dialysis Tubing, 8000 MWCO, 40mm/25.5mm; 16ft EW-02890-53 Mfr # 132116
COMPARE
25.5 40 8000
$332.00USD / Box of 1
Spectra Por S/P 7 7 RC Dialysis Tubing, 8000 MWCO, 50mm/32mm; 16ft EW-02890-54 Mfr # 132131
COMPARE
32 50 8000
$332.00USD / Box of 1
Spectra Por S/P 7 7 RC Dialysis Tubing, 10000 MWCO; diameter 15 mm; 16 ft (5 m) length. EW-02890-55 Mfr # 132118
COMPARE
15 24 10000
$332.00USD / Box of 1
Spectra Por S/P 7 7 RC Dialysis Tubing, 10000 MWCO, 45mm/29mm; 16ft EW-02890-56 Mfr # 132120
COMPARE
29 45 10000
$374.00USD / Box of 1
Spectra Por S/P 7 7 RC Dialysis Tubing, 15000 MWCO, 45mm/29mm; 16ft EW-02890-58 Mfr # 132124
COMPARE
29 45 15000
$332.00USD / Box of 1
Spectra Por S/P 7 7 RC Dialysis Tubing, 25000 MWCO, 34mm/22mm; 16ft EW-02890-59 Mfr # 132127
COMPARE
22 34 25000
$411.00USD / Box of 1
Spectra Por S/P 7 7 RC Dialysis Tubing, 50000 MWCO, 34mm/22mm; 16ft EW-02890-61 Mfr # 132130
COMPARE
21.6 34 50000
$374.00USD / Box of 1
Spectra Por S/P 7 7 Membranes; MWCO, 1000; flat width, 18mm; diameter, 11.5mm EW-02902-54 Mfr # 132103
COMPARE
11.5 18 1000
$374.00USD / Box of 1
Spectra Por S/P 7 7 Membranes; MWCO, 1000; flat width, 38mm; diameter, 24.2mm EW-02902-56 Mfr # 132104
COMPARE
24.2 38 1000
$374.00USD / Box of 1
Spectra Por S/P 7 7 Membranes; MWCO, 1000; flat width, 45mm; diameter, 28.6mm EW-02902-58 Mfr # 132105
COMPARE
28.6 45 1000
$411.00USD / Box of 1
Spectra Por S/P 7 7 Membranes; MWCO, 10,000; flath width, 12mm; diameter, 7.6mm EW-02902-60 Mfr # 132117
COMPARE
7.6 12 10000
$332.00USD / Box of 1
Spectra Por S/P 7 7 Membranes; MWCO, 10,000; flat width, 32mm, diameter, 20.4mm EW-02902-62 Mfr # 132119
COMPARE
20.4 32 10000
$374.00USD / Box of 1
Spectra Por S/P 7 7 Membranes; MWCO, 15,000; flat width, 28mm, diameter, 17.8mm EW-02902-66 Mfr # 132123
COMPARE
20.4 32 15000
$374.00USD / Box of 1
Showing 1 - 20 of 22