Spectra/Por 2 Regenerated Cellulose (RC) Dialysis Discs and Sheets

Available in a variety of dimensions to meet your applications needs
  • Regenerated Cellulose (RC) membrane for the gentle separation and purification of solutes
MWCO of 12,000 to 14,000 Dalton
$207.00 - $381.00USD / Pkg of 25

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max Molecular Weight Cut Off (Mwco) (Dalton) Min Molecular Weight Cut Off (Mwco) (Dalton) Description
Availability Pricing
Spectra Por S/P 2 2 RC Dialysis Membrane Disc, 12000-14000 MWCO, 33mm Dia; 50/Pk EW-02890-64 Mfr # 132482
COMPARE
14000 12000 2 RC Dialysis Membrane Disc, 12000-14000 MWCO, 33mm Dia; 50/Pk
$207.00USD / Pkg of 50
Spectra Por S/P 2 2 RC Dialysis Membrane Disc, 12000-14000 MWCO, 47mm Dia; 50/Pk EW-02890-65 Mfr # 132480
COMPARE
14000 12000 2 RC Dialysis Membrane Disc, 12000-14000 MWCO, 47mm Dia; 50/Pk
$250.00USD / Pkg of 50
Spectra Por S/P 2 Spectr/Por 2 Membranes Flat sheets; 240 x 240 mm; 25/pack EW-02900-24 Mfr # 132686
COMPARE
14000 12000 Spectr/Por 2 Membranes Flat sheets; 240 x 240 mm; 25/pack
$381.00USD / Pkg of 25
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat