Spectra/Mesh Screen Discs and Sheets
Four material option ensure chemical and application compatibility
  • Materials include polypropylene, nylon, PEEK, and 304 stainless steel
There are four material options to choose from. The nylon mesh filters out particles down to 8 µm. These are ideal for finer macrofiltration needs and are suitable with all aqueous solutions. Polypropylene (PP) mesh offers strength and stability, while also being resistant to most acids, alkalies, and alcohols. These screens are autoclavable. The PEEK mesh holds its shape and features superior chemical resistance. These are ideal for general filtration, are resistant to acidic and alkaline solutions, and feature temperature stability up to 250°C. The 304 Stainless Steel woven discs or sheets are ideal for high differential pressure applications as they are durable and feature extra strength.
$132.00 - $685.00USD / Pkg of 3
47 variations of this product are available.
See All Product Options Below
47 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Material Pore Size (µm)
Availability Pricing
Spectra/Mesh 146418 Screen Sheets, Polypropylene, 500 µm, 3/pk EW-06630-44 Mfr # 146418
COMPARE
300 Polypropylene 500
$241.00USD / Pkg of 3
Spectra/Mesh 146422 Screen Sheets, Polypropylene, 350 µm, 3/pk EW-06630-45 Mfr # 146422
COMPARE
300 Polypropylene 350
$241.00USD / Pkg of 3
Spectra/Mesh 146428 Screen Sheets, Polypropylene; pore size, 210µ; 30 x 30-cm sheets EW-06630-48 Mfr # 146428
COMPARE
300 Polypropylene 210
$241.00USD / Pkg of 3
Spectra/Mesh 146432 Screen Sheets, Polypropylene, 30 x 30 cm, 149 µm, 3/pk EW-06630-50 Mfr # 146432
COMPARE
300 Polypropylene 149
$241.00USD / Pkg of 3
Spectra/Mesh 146488 Screen Sheets, 30 X 30 cm Nylon, 100 µm, 3/pk EW-06630-75 Mfr # 146488
COMPARE
300 Nylon 100
$388.00USD / Pkg of 3
Spectra/Mesh 146490 Screen Sheets, 30 X 30 cm Nylon, 70 µm, 3/pk EW-06630-80 Mfr # 146490
COMPARE
300 Nylon 70
$388.00USD / Pkg of 3
Spectra/Mesh 146498 Screen Sheets, 30 X 30 cm Nylon, 52 µm, 3/pk EW-06630-84 Mfr # 146498
COMPARE
300 Nylon 53
$388.00USD / Pkg of 3
Spectra/Mesh 146502 Screen Sheets, 30 X 30 cm Nylon, 41 µm, 3/pk EW-06630-86 Mfr # 146502
COMPARE
300 Nylon 41
$388.00USD / Pkg of 3
Spectra/Mesh 146506 Screen Sheets, 30 X 30 cm Nylon, 30 µm, 3/pk EW-06630-88 Mfr # 146506
COMPARE
300 Nylon 30
$398.00USD / Pkg of 3
Spectra/Mesh 146510 Screen Sheets, 30 X 30 cm Nylon, 20 µm, 3/pk EW-06630-90 Mfr # 146510
COMPARE
300 Nylon 20
$398.00USD / Pkg of 3
Spectra/Mesh 146514 Screen Sheets, 30 X 30 cm Nylon, 10 µm, 3/pk EW-06630-92 Mfr # 146514
COMPARE
300 Nylon 10
$398.00USD / Pkg of 3
Spectra/Mesh 146439 Screen Sheets, Stainless Steel, 30 x 30 cm, 103 µm, 2/pk EW-06630-97 Mfr # 146439
COMPARE
300 Stainless Steel 103
$247.00USD / Pkg of 2
Spectra/Mesh 148130 Screen Discs, 47 mm, Nylon, 5 µm, pack of 10 EW-06631-00 Mfr # 148130
COMPARE
47 Nylon 5
$167.00USD / Pkg of 10
Spectra/Mesh 148132 Screen Discs, 47 mm, Nylon, 10 µm, pack of 10 EW-06631-02 Mfr # 148132
COMPARE
47 Nylon 10
$217.00USD / Pkg of 10
Spectra/Mesh 148134 Screen Discs, 47 mm, Nylon, 20 µm, pack of 10 EW-06631-03 Mfr # 148134
COMPARE
47 Nylon 20
$217.00USD / Pkg of 10
Spectra/Mesh 148136 Screen Discs, 47 mm, Nylon, 30 µm, pack of 10 EW-06631-04 Mfr # 148136
COMPARE
47 Nylon 30
$167.00USD / Pkg of 10
Spectra/Mesh 148138 Screen Discs, 47 mm, Nylon, 41 µm, pack of 10 EW-06631-05 Mfr # 148138
COMPARE
47 Nylon 41
$211.00USD / Pkg of 10
Spectra/Mesh 148140 Screen Discs, 47 mm, Nylon, 53 µm, pack of 10 EW-06631-06 Mfr # 148140
COMPARE
47 Nylon 53
$210.00USD / Pkg of 10
Spectra/Mesh 148143 Screen Discs, 47 mm, Nylon, 70 µm, pack of 10 EW-06631-08 Mfr # 148143
COMPARE
47 Nylon 70
$211.00USD / Pkg of 10
Spectra/Mesh 148145 Screen Discs, 47 mm, Nylon, 100 µm, pack of 10 EW-06631-09 Mfr # 148145
COMPARE
47 Nylon 100
$211.00USD / Pkg of 10
Showing 1 - 20 of 47