Special-Service Regulators
Model 03274-10 is a single-stage unit specifically designed for gases that attack brass or copper-bearing alloys. When used with low-vapor-pressure liquefied gases, no inlet gauge is needed. Rubber-supported PTFE diaphragm, KEL-F seat, aluminum-alloy body and bonnet, SS delivery gauge, and 10 mm sintered SS filter.
ITEM(S) ON SALE
$1,297.10 - $2,858.55USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (SCFH) CGA Fitting Max Inlet Pressure (PSI)
Availability Pricing
COMPARE
780 330 3000
$2,858.55USD / Each
$3,363.00USD / Each
COMPARE
240 350 3000
$1,297.10USD / Each
$1,526.00USD / Each