Acros Organics Sodium Carbonate

Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
The Acros Organics brand delivers consistent, high-quality organics and fine chemicals at great savings. It also focuses on the evolving trends in organic chemistry, supporting research, development and product activities through a range of off-the-shelf products and expertise in sourcing, manufacturing and technology partnerships. Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$112.00 - $199.50USD / Each

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Weight (g) Description
Availability Pricing
Acros Organics Sodium carbonate, anhydrous, pure (1kg) CAS 497-19-8 EW-88122-38 Mfr # AC123670010
COMPARE
1000 Sodium carbonate, anhydrous, pure (1kg) CAS 497-19-8
$134.75USD / Each
Acros Organics Sodium carbonate decahydrate p.a. (1kg) CAS 6132-02-1 EW-88228-03 Mfr # AC213500010
COMPARE
1000 Sodium carbonate decahydrate p.a. (1kg) CAS 6132-02-1
$112.00USD / Each
Acros Organics Sodium carbonate, anhydrous, powder, reagent ACS (500g) EW-88362-72 Mfr # AC424285000
COMPARE
500 Sodium carbonate, anhydrous, powder, reagent ACS (500g)
$199.50USD / Each
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat