SNOL Muffle Furnaces
Designed for a variety thermal processes up to 1300 °C.
  • One-piece chamber and ecological insulation provide high thermal efficiency
  • Low power consumption saves electricity
  • Short heating up/cooling down period for efficient process time
Using high quality thermoinsulation, ceramic hearth plates, and embedded heating elements on models up to 1100°C (exposed on ceramic tubes on models up to 1300°C) these furnaces provide excellent temperature stability and uniformity. Equipped with high-precision digital microprocessor controllers fitted with self-tuning and manual PID settings, they are an optimal fit for scientific, educational, government, industrial, and material testing labs.
$2,306.00 - $6,816.00USD / Each
19 variations of this product are available.
See All Product Options Below
19 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (Liters) Power (VAC) Description
Availability Pricing
SNOL 3/1100 LHM01  Muffle Furnace, 3 L, Upward Opening Door, 120V EW-33859-06 Mfr # SNOL 3/1100 LHM01 US
COMPARE
3 120 Muffle Furnace, 3 L, Upward Opening Door, 120V
$2,306.00USD / Each
SNOL 3/1100 LHM01  Muffle Furnace, 3 L, Upward Opening Door, 230V EW-33859-07 Mfr # SNOL 3/1100 LHM01 EU
COMPARE
3 230 Muffle Furnace, 3 L, Upward Opening Door, 230V
$2,306.00USD / Each
SNOL 8.2/1100 LHM01  Muffle Furnace, 8.2 , Upward Opening Door, 120V EW-33859-08 Mfr # SNOL 8.2/1100 LHM01 US
COMPARE
8.2 120 Muffle Furnace, 8.2 , Upward Opening Door, 120V
$2,705.00USD / Each
SNOL 8.2/1100 LHM01  Muffle Furnace, 8.2 L, Upward Opening Door, 230V EW-33859-09 Mfr # SNOL 8.2/1100 LHM01 EU
COMPARE
8.2 230 Muffle Furnace, 8.2 L, Upward Opening Door, 230V
$2,705.00USD / Each
SNOL 8.2/1100 LSM01  Muffle Furnace, 8.2 L, Sideways Opening Door, 120V EW-33859-14 Mfr # SNOL 8.2/1100 LSM01 US
COMPARE
8.2 120 Muffle Furnace, 8.2 L, Sideways Opening Door, 120V
$2,493.00USD / Each
SNOL 8.2/1100 LSM01  Muffle Furnace, 8.2 L, Sideways Opening Door, 230V EW-33859-15 Mfr # SNOL 8.2/1100 LSM01 EU
COMPARE
8.2 230 Muffle Furnace, 8.2 L, Sideways Opening Door, 230V
$2,387.00USD / Each
SNOL 13/1100 LHM01  Muffle Furnace, 13 L, Upwards Opening Door, 120V EW-33859-18 Mfr # SNOL 13/1100 LHM01 US
COMPARE
13 120 Muffle Furnace, 13 L, Upwards Opening Door, 120V
$3,485.00USD / Each
SNOL 13/1100 LHM01  Muffle Furnace, 13 L, Upwards Opening Door, 230V EW-33859-19 Mfr # SNOL 13/1100 LHM01 EU
COMPARE
13 230 Muffle Furnace, 13 L, Upwards Opening Door, 230V
$3,341.00USD / Each
SNOL 22/1100 LHM01  Muffle Furnace, 22 L, Upwards Opening Door, 120V EW-33859-20 Mfr # SNOL 22/1100 LHM01 US
COMPARE
22 120 Muffle Furnace, 22 L, Upwards Opening Door, 120V
$4,607.00USD / Each
SNOL 22/1100 LHM01  Muffle Furnace, 22 L, Upwards Opening Door, 230V EW-33859-21 Mfr # SNOL 22/1100 LHM01 EU
COMPARE
22 230 Muffle Furnace, 22 L, Upwards Opening Door, 230V
$4,426.00USD / Each
SNOL 39/1100 LHM01  Muffle Furnace, 39 L,  Upwards Opening Door, 230V EW-33859-22 Mfr # SNOL 39/1100 LHM01 US
COMPARE
39 230 Muffle Furnace, 39 L, Upwards Opening Door, 230V
$5,936.00USD / Each
SNOL 6.7/1300 LSM01  Muffle Furnace, 6.7 L, Sideways Opening Door, 120V EW-33859-24 Mfr # SNOL 6.7/1300 LSM01 US
COMPARE
6.7 120 Muffle Furnace, 6.7 L, Sideways Opening Door, 120V
$3,795.00USD / Each
SNOL 6.7/1300 LSM01  Muffle Furnace, 6.7 L, Sideways Opening Door, 230V EW-33859-25 Mfr # SNOL 6.7/1300 LSM01 EU
COMPARE
6.7 230 Muffle Furnace, 6.7 L, Sideways Opening Door, 230V
$3,795.00USD / Each
SNOL 3/1100 LHM01 SS  Muffle Furnace, Stainless Steel, 3 L, Upwards Opening Door, 230V EW-33859-27 Mfr # SNOL 3/1100 LHM01 SS EU
COMPARE
3 230 Muffle Furnace, Stainless Steel, 3 L, Upwards Opening Door, 230V
$2,529.00USD / Each
SNOL 8.2/1100 LHM01 SS  Muffle Furnace, Stainless Steel, 8.2 L, Upwards Opening Door, 120V EW-33859-28 Mfr # SNOL 8.2/1100 LHM01 SS US
COMPARE
8.2 120 Muffle Furnace, Stainless Steel, 8.2 L, Upwards Opening Door, 120V
$3,250.00USD / Each
SNOL 3/1100 LHM01 SS  Muffle Furnace, Stainless Steel, 13 L, Upwards Opening Door, 120V EW-33859-30 Mfr # SNOL 13/1100 LHM01 SS US
COMPARE
13 120 Muffle Furnace, Stainless Steel, 13 L, Upwards Opening Door, 120V
$4,426.00USD / Each
SNOL 3/1100 LHM01 SS  Muffle Furnace, Stainless Steel, 13 L, Upwards Opening Door, 230V EW-33859-31 Mfr # SNOL 13/1100 LHM01 SS EU
COMPARE
13 230 Muffle Furnace, Stainless Steel, 13 L, Upwards Opening Door, 230V
$4,426.00USD / Each
SNOL 22/1100 LHM01 SS  Muffle Furnace, Stainless Steel, 22 L, Upwards Opening Door, 230V EW-33859-33 Mfr # SNOL 22/1100 LHM01 SS EU
COMPARE
22 230 Muffle Furnace, Stainless Steel, 22 L, Upwards Opening Door, 230V
$5,508.00USD / Each
SNOL 39/1100 LHM01 SS  Muffle Furnace, Stainless Steel, 39 L, Upwards Opening Door, 230V EW-33859-34 Mfr # SNOL 39/1100 LHM01 SS US
COMPARE
39 230 Muffle Furnace, Stainless Steel, 39 L, Upwards Opening Door, 230V
$6,816.00USD / Each
Showing 1- 19 of 19