Small Sample Adapters for Brookfield Rotational Viscometers

Use the small sample adapters when measuring low fluid volumes of 8 to 13 mL. Adapters include a water jacket for attachment to an external circulating bath (not included). Easily install or remove sample chamber without disturbing operation of viscometer or circulating bath. Adapters operate from 5 to 212°F (-15 to 100°100).
$1,942.00 - $2,229.00USD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Viscosity Range (cP)
Availability Pricing
Brookfield AMETEK SSA18/13R Small sample adaptor for models 98945-00 and -05; 3 to 10 K cp EW-98945-56 Mfr # SSA18/13R
COMPARE
10000
$2,176.00USD / Each
Brookfield AMETEK SSA31/13R Small sample adapter for models 98945-00 and -05; 30 to 100 K cp EW-98945-58 Mfr # SSA31/13R
COMPARE
100000
$1,942.00USD / Each
Brookfield AMETEK SSA34/13R Small sample adapter for models 98945-00 and -05; 60 to 200 K cp EW-98945-60 Mfr # SSA34/13R
COMPARE
200000
$2,022.00USD / Each
Brookfield AMETEK SSA27/13R Small Sample Adapter; 250-830K (Med)/2K-6.7M (High) EW-98945-64 Mfr # SSA27/13R
COMPARE
6700000
$2,229.00USD / Each
Brookfield AMETEK SSA28/13R Small Sample Adapter; 500-1.7M (Med)/4K-13.3M (High) EW-98945-66 Mfr # SSA28/13R
COMPARE
13300000
$2,195.00USD / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat