This product is now discontinued, View Related Products
SHUR/Mount Liquid Coverglass and Mounting Medium
Mount and preserve your microscope slide specimens
  • Convenient twisty spout eliminates messy bottles and droppers
  • Anti-oxidant prevents fading and UV absorbent prevents yellowing
  • Low viscosity minimizes bubble formation under coverglass
Use the toluene-based acrylic resin for rapid drying and hand coverslipping. Use the xylene-based mounting medium for automated coverslipping instruments. Water-based mounting medium is for immuno procedures.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Description
Availability Pricing
TBS LC-A /Mount Low Viscosity Mounting Medium, Xylene-Based; 16 oz EW-48579-05 Mfr # LC-A
COMPARE
473 /Mount Low Viscosity Mounting Medium, Xylene-Based; 16 oz
Call For Price
COMPARE
SHUR/Mount™ Mounting medium, Water based for immuno procedures; 1oz (20ml)/ea